siRNA Details
Gene Symbol: Tmem29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164686.1 | NP_001158158.1 | 3 | 336 | ||
GenBank mRNA | AK012546.1 | - | 3 | 347 | ||
BC058428.1 | - | 3 | 297 | |||
CA526769.1 | - | 3 | 337 | |||
CK624690.1 | - | 3 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
