siRNA Details
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 5 | 293 | ||
GenBank mRNA | BC049626.1 | - | 5 | 293 | 6705850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
