siRNA Details
Gene Symbol: Tmed8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033475.3 | NP_001028647.1 | 3 | 267 | ||
GenBank mRNA | AK147378.1 | - | 3 | 268 | ||
BC150860.1 | - | 3 | 338 | 9087771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
