siRNA Details
Gene Symbol: Cbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007624.3 | NP_031650.3 | 3 | 239 | ||
GenBank mRNA | AK002910.1 | - | 3 | 214 | ||
AK083957.1 | - | 3 | 154 | |||
AK146376.1 | - | 3 | 189 | |||
BC003840.1 | - | 3 | 166 | |||
BC018354.1 | - | 2 | 121 | 3990224 | ||
BC059831.1 | - | 3 | 142 | 5717629 | ||
BC108370.1 | - | 3 | 166 | 30728322 | ||
BC110376.1 | - | 3 | 196 | 3467208 | ||
CJ093936.1 | - | 3 | 240 | |||
X56683.1 | - | 1 | 56 |

More Information
Other Species Targeted

Gene Ontology Categories:
