siRNA Details
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 307 | ||
GenBank mRNA | BC089025.1 | - | 2 | 307 | 30618054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
