siRNA Details
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027166.5 | NP_081442.1 | 2, 3 | 262 | ||
GenBank mRNA | AB098744.1 | - | 1, 2 | 150 | ||
AK010201.1 | - | 2, 3 | 228 | |||
AK046281.1 | - | 2, 3 | 261 | |||
AK049096.1 | - | 2, 3 | 263 | |||
AK049644.1 | - | 2, 3 | 261 | |||
AK137446.1 | - | 2, 3 | 234 | |||
AK154461.1 | - | 2, 3 | 301 | |||
AK165943.1 | - | 2, 3 | 264 | |||
AK167998.1 | - | 2, 3 | 278 | |||
AK169241.1 | - | 2, 3 | 265 | |||
AK169986.1 | - | 2, 3 | 236 | |||
AK171010.1 | - | 2, 3 | 263 | |||
AK217637.1 | - | 1, 2 | 176 | |||
BC025125.1 | - | 1 | 22 | 4921751 | ||
BC085109.1 | - | 2, 3 | 281 | 30737658 |

More Information
Other Species Targeted

Gene Ontology Categories:
