siRNA Details
Gene Symbol: Pramel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085418.2 | NP_001078887.1 | 3 | 591 | ||
GenBank mRNA | BC052827.1 | - | 3 | 601 | 30033377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
