siRNA Details
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011119.3 | NP_035249.1 | 10 | 1136 | ||
GenBank mRNA | AK002685.1 | - | 10 | 1213 | ||
AK076286.1 | - | 10 | 1142 | |||
AK144645.1 | - | 10 | 1190 | |||
AK166936.1 | - | 10 | 1145 | |||
AK168582.1 | - | 10 | 1138 | |||
BC003777.1 | - | 10 | 1126 | |||
BC005593.1 | - | 10 | 1126 | |||
BC028926.1 | - | 10 | 1119 | |||
BC037623.1 | - | 10 | 1156 | |||
BC046532.1 | - | 10 | 1090 | 5358060 | ||
BC086788.1 | - | 8 | 3342 | |||
U43918.1 | - | 10 | 1295 | |||
X84789.1 | - | 10 | 1201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
