siRNA Details
Gene Symbol: Gatc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029645.3 | NP_083921.1 | 2 | 397 | ||
GenBank mRNA | AK008080.1 | - | 2 | 402 | ||
AK034326.1 | - | 2 | 397 | |||
BC021775.1 | - | 2 | 390 | 4158979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
