siRNA Details
Gene Symbol: Gm5382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 300 | ||
GenBank mRNA | BC100482.1 | - | 1 | 300 | 30918325 |
Gene Symbol: Gm14474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242947.1 | NP_001229876.1 | 1 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
