siRNA Details
Gene Symbol: Thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146153.3 | NP_666265.3 | 4 | 1318 | ||
GenBank mRNA | AK034044.1 | - | 4 | 1198 | ||
AK079601.1 | - | 4 | 1198 | |||
BC040346.1 | - | 4 | 1249 | |||
BC092239.1 | - | 4 | 1234 | 30659321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
