siRNA Details
Gene Symbol: Lca5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001492.2 | NP_001001492.2 | 7 | 1336 | ||
GenBank mRNA | AK029554.1 | - | 7 | 1330 | ||
AK133316.1 | - | 7 | 1337 | |||
BC141285.1 | - | 6 | 1186 | 9055897 | ||
BC145611.1 | - | 6 | 1186 | 9054177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
