siRNA Details
Gene Symbol: Zfp174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081217.1 | NP_001074686.1 | 3 | 1491 | ||
GenBank mRNA | AK141897.1 | - | 3 | 1429 | ||
BC079601.1 | - | 3 | 1401 | |||
BC147582.1 | - | 3 | 1446 | 9056572 | ||
BC151072.1 | - | 3 | 1445 | 9087985 | ||
BC151078.1 | - | 3 | 1446 | 9087991 |

More Information
Other Species Targeted

Gene Ontology Categories:
