siRNA Details
Gene Symbol: Zfp174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081217.1 | NP_001074686.1 | 2 | 1202 | ||
GenBank mRNA | AK141897.1 | - | 2 | 1140 | ||
BC079601.1 | - | 2 | 1112 | |||
BC147582.1 | - | 2 | 1157 | 9056572 | ||
BC151072.1 | - | 2 | 1156 | 9087985 | ||
BC151078.1 | - | 2 | 1157 | 9087991 |
