siRNA Details
Gene Symbol: Eid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198425.2 | NP_940817.1 | 1 | 736 | ||
GenBank mRNA | AY251273.1 | - | 1 | 662 | ||
BC094513.1 | - | 1 | 736 | 2648407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
