siRNA Details
Gene Symbol: Tas2r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199154.1 | NP_954605.1 | 1 | 506 | ||
GenBank mRNA | AF412301.1 | - | 1 | 341 | ||
BC141027.1 | - | 1 | 571 | 9055639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
