siRNA Details
Gene Symbol: Ssxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026492.3 | NP_080768.1 | 7 | 646 | ||
GenBank mRNA | AK006218.1 | - | 7 | 647 | ||
AK015135.1 | - | 7 | 647 | |||
AY244792.1 | - | 7 | 647 | |||
BC145538.1 | - | 7 | 619 | 9054101 | ||
BC145541.1 | - | 564 | 9054104 | |||
BC145782.1 | - | 7 | 538 | 40131085 | ||
BC145784.1 | - | 7 | 538 | 40131087 | ||
BC171964.1 | - | 619 | 9054250 |
Gene Symbol: Gm14459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126491.1 | NP_001119963.1 | 6 | 492 |
Gene Symbol: Gm6592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081564.1 | NP_001075033.1 | 6 | 459 |
Gene Symbol: Ssx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199063.2 | NP_951018.1 | 6 | 511 | ||
GenBank mRNA | AY244795.1 | - | 6 | 511 |
Gene Symbol: Ssxb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199064.1 | NP_951019.1 | 7 | 647 | ||
GenBank mRNA | AY244801.1 | - | 7 | 647 | ||
BC145539.1 | - | 619 | 9054102 |
Gene Symbol: Ssxb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244802.1 | - | 6 | 510 |
Gene Symbol: Ssxb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244803.1 | - | 6 | 510 |
Gene Symbol: Ssxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198898.2 | NP_942601.1 | 6 | 511 | ||
GenBank mRNA | AY244794.1 | - | 6 | 511 | ||
BC087939.1 | - | 7 | 591 | |||
BC132618.1 | - | 6 | 511 | 40130895 | ||
BC132644.1 | - | 6 | 511 | 40130921 |
Gene Symbol: Ssxb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199319.3 | NP_955023.1 | 6 | 510 | ||
GenBank mRNA | AY244796.1 | - | 6 | 510 |
Gene Symbol: Ssxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205108.1 | NP_001192037.1 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
