siRNA Details
Gene Symbol: Opa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207525.3 | NP_997408.2 | 2 | 226 | ||
GenBank mRNA | AK133480.1 | - | 212 | |||
AK154917.1 | - | 232 | ||||
AK154960.1 | - | 227 | ||||
AK164755.1 | - | 2 | 227 | |||
AK165767.1 | - | 2 | 257 | |||
AK170977.1 | - | 2 | 275 | |||
BC094601.1 | - | 2 | 262 | 6404450 | ||
BC132309.1 | - | 2 | 211 | 40130586 | ||
BC132311.1 | - | 2 | 211 | 40130588 |
