siRNA Details
Gene Symbol: Mrgprb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207537.1 | NP_997420.1 | 1 | 124 | ||
GenBank mRNA | BC107192.1 | - | 1 | 123 | ||
BC107193.1 | - | 1 | 123 | |||
BC114400.1 | - | 1 | 123 | |||
BC114428.1 | - | 1 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
