siRNA Details
Gene Symbol: Mrgprb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207538.1 | NP_997421.1 | 1 | 61 | ||
GenBank mRNA | BC120849.1 | - | 1 | 94 | 40129772 | |
BC132003.1 | - | 1 | 95 | 40130280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
