siRNA Details
Gene Symbol: Olfr192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207549.1 | NP_997432.1 | 1 | 238 |
Gene Symbol: Olfr190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146397.2 | NP_666509.2 | 1 | 238 | ||
GenBank mRNA | BC104050.1 | - | 1 | 238 | 40040225 | |
BC104051.1 | - | 1 | 238 | 40040226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
