siRNA Details
Gene Symbol: Olfr192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207549.1 | NP_997432.1 | 1 | 515 |
Gene Symbol: Olfr190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146397.2 | NP_666509.2 | 1 | 515 | ||
GenBank mRNA | BC104050.1 | - | 1 | 515 | 40040225 | |
BC104051.1 | - | 1 | 515 | 40040226 |

More Information
Other Species Targeted

Gene Ontology Categories:
