siRNA Details
Gene Symbol: Olfr403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207622.1 | NP_997505.1 | 1 | 454 | ||
GenBank mRNA | AF102517.1 | - | 1 | 265 | ||
BC104289.1 | - | 1 | 454 | 40042769 | ||
BC104290.1 | - | 1 | 454 | 40042770 |
Gene Symbol: Olfr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146711.2 | NP_666922.2 | 1 | 529 | ||
GenBank mRNA | BC125464.1 | - | 1 | 529 | 40129979 | |
BC125468.1 | - | 1 | 529 | 40129983 |
