siRNA Details
Gene Symbol: Olfr750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207558.2 | NP_997441.2 | 1 | 947 | ||
GenBank mRNA | BC145881.1 | - | 1 | 947 | 40131184 |

More Information
Other Species Targeted

Gene Ontology Categories:
