siRNA Details
Gene Symbol: Olfr1335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207703.1 | NP_997586.1 | 1 | 329 | ||
GenBank mRNA | BC147439.1 | - | 1 | 411 | 9056081 | |
BC147453.1 | - | 1 | 411 | 9056095 |
Gene Symbol: Olfr1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146399.2 | NP_666511.2 | 1 | 275 | ||
GenBank mRNA | BC106978.2 | - | 1 | 275 | 40042180 | |
BC148290.2 | - | 1 | 275 | 40042169 |
Gene Symbol: Olfr1329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011870.2 | NP_001011870.2 | 1 | 275 |
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 281 | ||
GenBank mRNA | BC108943.1 | - | 1 | 281 | 40042936 | |
BC108944.1 | - | 1 | 276 |
Gene Symbol: Olfr1337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146309.3 | NP_666421.2 | 1 | 284 | ||
GenBank mRNA | BC127989.1 | - | 1 | 280 | 40135644 | |
BC152318.1 | - | 1 | 286 |
Gene Symbol: Olfr1338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207152.2 | NP_997035.2 | 1 | 275 | ||
GenBank mRNA | BC130245.1 | - | 1 | 279 | 40142324 |

More Information
Other Species Targeted

Gene Ontology Categories:
