siRNA Details
Gene Symbol: Olfr1380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207573.1 | NP_997456.1 | 1 | 67 | ||
GenBank mRNA | BC106971.1 | - | 1 | 67 | 40041445 | |
BC106972.1 | - | 1 | 67 | 40041448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
