siRNA Details
Gene Symbol: Olfr1383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207574.1 | NP_997457.1 | 1 | 97 | ||
GenBank mRNA | BC127965.1 | - | 1 | 101 | 40134985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
