siRNA Details
Gene Symbol: Olfr221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001808.2 | NP_001001808.2 | 1 | 406 | ||
GenBank mRNA | BC147427.1 | - | 1 | 440 | 9056495 | |
BC147428.1 | - | 1 | 440 | 9056478 | ||
BC151091.1 | - | 1 | 712 | 9088004 | ||
BC151095.1 | - | 1 | 705 | 9088008 |
