siRNA Details
Gene Symbol: Bex4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212457.2 | NP_997622.1 | 3 | 409 | ||
GenBank mRNA | AK010400.1 | - | 3 | 412 | ||
AY833558.1 | - | 3 | 409 | |||
BC090663.1 | - | 3 | 386 | 30677421 | ||
BC116226.2 | - | 3 | 313 | 40091511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
