siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199083.1 | NP_001186012.1 | 10 | 4245 | ||
NM_001199084.1 | NP_001186013.1 | 9 | 3987 | |||
GenBank mRNA | BC146009.1 | - | 1 | 923 | 40131312 | |
BC146011.1 | - | 1 | 923 | 40131314 | ||
CN693598.1 | - | 1 | 120 |

More Information
Other Species Targeted

Gene Ontology Categories:
