siRNA Details
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 5 | 723 | ||
GenBank mRNA | AK080983.1 | - | 864 | |||
AK088196.1 | - | 5 | 736 | |||
AK151326.1 | - | 709 | ||||
AK151705.1 | - | 5 | 733 | |||
BC002270.1 | - | 5 | 723 | 3496088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
