siRNA Details
Gene Symbol: Ndufs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010888.2 | NP_035018.1 | 3, 4 | 313 | ||
GenBank mRNA | AK147147.1 | - | 3, 4 | 343 | ||
BC059730.1 | - | 3, 4 | 286 | |||
BC076501.1 | - | 3, 4 | 344 | |||
BC086933.1 | - | 3, 4 | 286 | 6760537 | ||
BG081040.2 | - | 3, 4 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
