siRNA Details
Gene Symbol: Zfp941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001180.2 | NP_001001180.2 | 3 | 664 | ||
GenBank mRNA | AK045889.1 | - | 3 | 643 | ||
AK141045.1 | - | 642 | ||||
BC066028.1 | - | 3 | 618 | 5702088 | ||
BC068250.1 | - | 3 | 634 | 30018777 |
