siRNA Details
Gene Symbol: Zfp941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001180.2 | NP_001001180.2 | 3 | 1840 | ||
GenBank mRNA | AK141045.1 | - | 1818 | |||
BC066028.1 | - | 3 | 1794 | 5702088 | ||
BC068250.1 | - | 3 | 1810 | 30018777 |
