siRNA Details
Gene Symbol: BC053393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025435.3 | NP_001020606.2 | 4, 5 | 676 | ||
GenBank mRNA | AK140690.1 | - | 4, 5 | 531 | ||
AK166706.1 | - | 4, 5 | 553 | |||
AK166838.1 | - | 4, 5 | 553 | |||
BC052651.1 | - | 4, 5 | 667 | 30034019 | ||
BC053393.1 | - | 2860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
