siRNA Details
Gene Symbol: BC031181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001181.3 | NP_001001181.1 | 2 | 360 | ||
GenBank mRNA | AK031999.1 | - | 1 | 34 | ||
AK077642.1 | - | 2 | 293 | |||
AK131658.1 | - | 2 | 357 | |||
BC016084.1 | - | 2 | 360 | 4501468 | ||
BC027819.1 | - | 2 | 374 | 5320851 | ||
BC031181.1 | - | 2 | 355 | 4988367 |
