siRNA Details
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080924.2 | NP_001074393.1 | 3 | 625 | ||
NM_001290501.1 | NP_001277430.1 | 4 | 472 | |||
GenBank mRNA | AK133342.1 | - | 4 | 473 | ||
AK163639.1 | - | 3 | 625 | |||
BC151080.1 | - | 4 | 346 | 9087993 | ||
BC151083.1 | - | 4 | 346 | 9087996 |
