siRNA Details
Gene Symbol: Zfp456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001186.3 | NP_001001186.3 | 4 | 1278 | ||
GenBank mRNA | AK164227.1 | - | 4 | 1280 | ||
BC141197.1 | - | 4 | 1261 | 9055809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
