siRNA Details
Gene Symbol: Otud6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163191.1 | NP_001156663.1 | 1 | 94 | ||
GenBank mRNA | BC153817.1 | - | 1 | 94 | 40050367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
