siRNA Details
Gene Symbol: 4930404N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014836.3 | NP_001014836.2 | 3, 4 | 485 | ||
GenBank mRNA | AK015088.1 | - | 3, 4 | 456 | ||
BC139256.1 | - | 3, 4 | 481 | 8862278 | ||
BC145589.1 | - | 2, 3 | 555 | |||
BC145960.1 | - | 3, 4 | 481 | 40131263 | ||
BU945663.1 | - | 3, 4 | 488 |
