siRNA Details
Gene Symbol: Gnptab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004164.2 | NP_001004164.2 | 16 | 3167 | ||
GenBank mRNA | AK140867.1 | - | 15 | 3271 | ||
AK154710.1 | - | 8 | 2066 | |||
AK173132.1 | - | 16 | 3167 | |||
BC060638.1 | - | 18 | 2651 | 6849405 | ||
L36434.1 | - | 4 | 1233 |
