siRNA Details
Gene Symbol: Dnaic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034878.2 | NP_001030050.2 | 8 | 1291 | ||
GenBank mRNA | AK081229.1 | - | 5 | 595 | ||
BM950889.1 | - | 6 | 595 | |||
EF565270.1 | - | 7 | 907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
