siRNA Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 8, 9 | 895 | ||
GenBank mRNA | AK133061.1 | - | 8, 9 | 910 | ||
AK156138.1 | - | 9, 10 | 1047 | |||
AK157892.1 | - | 7, 8 | 1722 | |||
AK162620.1 | - | 8, 9 | 894 | |||
AK171660.1 | - | 5, 6 | 558 | |||
AK172423.1 | - | 8, 9 | 896 | |||
BC145584.1 | - | 8, 9 | 908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
