siRNA Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 9 | 903 | ||
GenBank mRNA | AK133061.1 | - | 9 | 918 | ||
AK156138.1 | - | 10 | 1055 | |||
AK157892.1 | - | 8 | 1730 | |||
AK162620.1 | - | 9 | 902 | |||
AK171660.1 | - | 6 | 566 | |||
AK172423.1 | - | 9 | 904 | |||
BC145584.1 | - | 9 | 916 |
