siRNA Details
Gene Symbol: LOC102642252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015882.1 | - | 1 | 144 | ||
AB774779.1 | - | 1 | 144 | |||
AB774832.1 | - | 1 | 144 | |||
AB774854.1 | - | 1 | 143 | |||
AF023206.1 | - | 1 | 106 | |||
AF023207.1 | - | 1 | 98 | |||
AF023227.1 | - | 1 | 66 | |||
AF072778.1 | - | 1 | 135 | |||
AF118998.1 | - | 1 | 120 | |||
AF276282.1 | - | 147 | ||||
AF303876.1 | - | 1 | 144 | |||
AF316525.1 | - | 1 | 135 | |||
AF547093.1 | - | 1 | 120 | |||
AY171930.1 | - | 1 | 120 | |||
AY171956.1 | - | 1 | 120 | |||
AY172509.1 | - | 120 | ||||
AY172529.1 | - | 120 | ||||
AY172544.1 | - | 1 | 120 | |||
AY172895.1 | - | 1 | 120 | |||
AY174003.1 | - | 1 | 120 | |||
AY182470.1 | - | 1 | 105 | |||
AY369883.1 | - | 1 | 114 | |||
AY594681.1 | - | 1 | 144 | |||
EF680939.1 | - | 1 | 125 | |||
EF680971.1 | - | 120 | ||||
EF680972.1 | - | 117 | ||||
EF680973.1 | - | 123 | ||||
EF680974.1 | - | 1 | 120 | |||
EF680975.1 | - | 117 | ||||
EF680976.1 | - | 120 | ||||
EU009128.1 | - | 141 | ||||
EU239771.1 | - | 1 | 121 | |||
EU915625.1 | - | 1 | 70 | |||
EU915639.1 | - | 1 | 70 | |||
EU934881.1 | - | 1 | 121 | |||
FJ816464.1 | - | 69 | ||||
FJ816492.1 | - | 69 | ||||
FJ816534.1 | - | 1 | 69 | |||
FJ816546.1 | - | 69 | ||||
FJ816550.1 | - | 69 | ||||
FJ816558.1 | - | 1 | 69 | |||
FJ816570.1 | - | 1 | 69 | |||
FJ816573.1 | - | 69 | ||||
FJ816595.1 | - | 1 | 69 | |||
FJ816599.1 | - | 69 | ||||
GU126674.1 | - | 1 | 144 | |||
HF569536.1 | - | 1 | 121 | |||
HF569540.1 | - | 1 | 121 | |||
HF569662.1 | - | 1 | 128 | |||
HF569693.1 | - | 1 | 121 | |||
HQ015075.1 | - | 1 | 144 | |||
HQ446591.1 | - | 1 | 123 | |||
HQ446592.1 | - | 1 | 117 | |||
HQ446628.1 | - | 1 | 117 | |||
HQ446629.1 | - | 1 | 131 | |||
HQ446630.1 | - | 1 | 117 | |||
HQ446649.1 | - | 123 | ||||
JX492330.1 | - | 1 | 129 | |||
JX492360.1 | - | 1 | 144 | |||
JX492388.1 | - | 1 | 144 | |||
JX492495.1 | - | 1 | 123 | |||
JX657377.1 | - | 1 | 121 | |||
JX657442.1 | - | 1 | 121 | |||
JX657516.1 | - | 1 | 121 | |||
JX657517.1 | - | 1 | 121 | |||
KC906652.1 | - | 1 | 144 | |||
KF207999.1 | - | 1 | 120 | |||
KF208007.1 | - | 1 | 120 | |||
KF208021.1 | - | 1 | 120 | |||
KF208066.1 | - | 1 | 120 | |||
KF208193.1 | - | 1 | 120 | |||
KF208218.1 | - | 120 | ||||
M36225.1 | - | 1 | 144 | |||
U55463.1 | - | 1 | 139 | |||
U55521.1 | - | 2 | 135 |
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154880.1 | - | 1 | 120 | ||
AF154909.1 | - | 1 | 120 | |||
AF154910.1 | - | 1 | 120 | |||
X73076.1 | - | 1 | 106 |
Gene Symbol: Ighg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092049.1 | - | 2 | 239 | 3661511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
