siRNA Details
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146238.3 | NP_666350.2 | 4 | 603 | ||
GenBank mRNA | AK037580.1 | - | 4 | 500 | ||
AK047511.1 | - | 5 | 698 | |||
AK053774.1 | - | 4 | 567 | |||
AK160583.1 | - | 4 | 604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
