siRNA Details
Gene Symbol: Uqcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026219.1 | NP_080495.1 | 3 | 243 | ||
GenBank mRNA | AK008455.1 | - | 3 | 219 | ||
AK008945.1 | - | 3 | 243 | |||
AK018769.1 | - | 3 | 219 | |||
AK168361.1 | - | 3 | 219 | |||
AK168855.1 | - | 3 | 234 | |||
BC031607.1 | - | 3 | 234 | 4460212 | ||
BC086921.1 | - | 3 | 218 | 6763146 | ||
BC100601.1 | - | 3 | 218 | 30121032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
