siRNA Details
Gene Symbol: Gm5464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034881.3 | NP_001030053.1 | 1 | 708 | ||
GenBank mRNA | AK135012.1 | - | 1 | 709 | ||
BC147346.1 | - | 1 | 706 | 9056391 | ||
BC147347.1 | - | 1 | 706 | 9056397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
