siRNA Details
Gene Symbol: Otulin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013792.2 | NP_001013814.2 | 4, 5 | 550 | ||
GenBank mRNA | AK134955.1 | - | 4, 5 | 385 | ||
AK150371.1 | - | 4, 5 | 553 | |||
BC087945.1 | - | 1, 2 | 151 | 30505408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
