siRNA Details
Gene Symbol: Eif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011508.1 | NP_035638.1 | 3 | 360 | ||
GenBank mRNA | AF129888.1 | - | 3 | 345 | ||
AK148032.1 | - | 3 | 357 | |||
AK150813.1 | - | 3 | 359 | |||
AK151070.1 | - | 3 | 357 | |||
AK151575.1 | - | 3 | 359 | |||
AK152194.1 | - | 3 | 359 | |||
AK169763.1 | - | 3 | 358 | |||
BC003463.1 | - | 3 | 360 | 2648657 | ||
BC010791.1 | - | 3 | 338 | 4218816 | ||
BC081429.1 | - | 3 | 347 | 5710951 | ||
BC096656.1 | - | 3 | 335 | 5143414 | ||
BC096691.1 | - | 3 | 345 | 5256396 | ||
Z50159.1 | - | 2 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
